Learn More
Hoefer™ SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarine Combs
Accessories for SUB20 Maxi-Standard and SUB20C Maxi-Cooled Submarines
845.00 NOK - 880.00 NOK
Specifications
For Use With (Equipment) | SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine |
---|---|
Type | Comb |
Depth (English) Pan | 0.19 in. |
Depth (Metric) Pan | 0.5 cm |
Product Code | Brand | Thickness (English) | Thickness (Metric) | No. of Wells | Width (Metric) Well | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (English) | Thickness (Metric) | No. of Wells | Width (Metric) Well | Price | Quantity & Availability | ||
12675766
|
Hoefer™
SUB20-C1.5-16 |
0.05 in. | 0.15 cm | 16 | 0.85 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12685766
|
Hoefer™
SUB20-C1.5-20MC |
0.05 in. | 0.15 cm | 20 | 0.7 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12695766
|
Hoefer™
SUB20-C1.5-28 |
0.05 in. | 0.15 cm | 28 | 0.5 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12605776
|
Hoefer™
SUB20-C1.5-40MC |
0.05 in. | 0.15 cm | 40 | 0.3 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12615776
|
Hoefer™
SUB20-C1-16 |
0.03 in. | 0.1 cm | 16 | 0.85 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12625776
|
Hoefer™
SUB20-C1-20MC |
0.03 in. | 0.1 cm | 20 | 0.7 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12635776
|
Hoefer™
SUB20-C1-28 |
0.03 in. | 0.1 cm | 28 | 0.5 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12645776
|
Hoefer™
SUB20-C1-40MC |
0.03 in. | 0.1 cm | 40 | 0.3 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12655776
|
Hoefer™
SUB20-C2-16 |
0.07 in. | 0.2 cm | 20 | 0.85 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12665776
|
Hoefer™
SUB20-C2-20MC |
0.07 in. | 0.2 cm | 20 | 0.7 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12675776
|
Hoefer™
SUB20-C2-28 |
0.07 in. | 0.2 cm | 28 | 0.5 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12685776
|
Hoefer™
SUB20-C2-40MC |
0.07 in. | 0.2 cm | 40 | 0.3 cm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Ranges from 16, 20, 28 and 40 Wells
- 1, 1.5 and 2 mm Thickness
- 3, 5, 7 and 8.5 mm Well Width
- Min. 13, max. 75 μL Sample Volume
Specifications
SUB20 Maxi Standard and SUB20C Maxi Cooled Submarine | |
Comb | |
0.19 in. | |
0.5 cm |