Learn More
Hoefer™ Combs for SUB25 Maxi-Plus Submarine Gel Electrophoresis Unit
For use with SUB25 and SUB25C Units
870.00 NOK
Specifications
For Use With (Application) | 52 sample throughput (multi-channel pipette compatible) |
---|---|
For Use With (Equipment) | SUB25 Maxi-Plus Unit |
Width (English) Gel | 9.84 in. |
Product Code | Brand | Thickness (English) | Thickness (Metric) | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (English) | Thickness (Metric) | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||
12685786
|
Hoefer™
SUB25-C1.5-52MC |
0.05 in. | 1.5 mm | 0.0006 oz. | 20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12605796
|
Hoefer™
SUB25-C1-52MC |
0.03 in. | 1 mm | 0.0004 oz. | 13 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12625796
|
Hoefer™
SUB25-C2-52MC |
0.07 in. | 2 mm | 0.0008 oz. | 25 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12675786
|
Hoefer™
SUB25-C1.5-26MC |
0.05 in. | 1.5 mm | 0.001 oz. | 45 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Either 26 or 52 Wells
- 1, 1.5 and 2 mm Thickness
- 3 or 7 mm Well Width
- Min. 13, max. 60 μL Sample Volume
Specifications
52 sample throughput (multi-channel pipette compatible) | |
SUB25 Maxi-Plus Unit | |
9.84 in. |