missing translation for 'onlineSavingsMsg'
Learn More

HNRNPD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12770-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16890664

  • 1281.92 NOK / 20µL
Estimated Shipment: 16-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

hnRNP D/AUF1 binds with high affinity to RNA molecules that contain AU-rich elements (AREs) found within the 3′ untranslated regions of many protooncogenes and cytokine mRNAs. It also binds to double and single stranded DNA sequences in a specific manner and functions a transcription factor. Each of the RNA-binding domains specifically can bind solely to a single stranded nonmonotonous 5′-UUAG-3′ sequence and also weaker to the single-stranded 5′-TTAGGG-3′ telomeric DNA repeat. It may be involved in translationally coupled mRNA turnover (referenced from swissprot).
Specifications

Specifications

HNRNPD
Polyclonal
Unconjugated
HNRNPD
AUF1, AUF1A, hnRNP D0, HNRNPD, hnRNPD0, HNRPD, P37
Rabbit
Antigen Affinity Chromatography
RUO
11991, 3184, 79256
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14103, Q60668, Q9JJ54
HNRNPD
HNRNPD Fusion Protein Ag3458
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers