missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His Tag Antibody (RMH01) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26048JF669
This item is not returnable.
View return policy
Description
His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for ELISA,Western Blot,Simple Western,Sandwich ELISA Capture,Sandwich ELISA Detection
Specifications
| His Tag | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 6 His epitope tag, 6-His Tag, 6X His, 6X His Tag, 6X-His, 6x-His Tag, H, Hexa His tag, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, His6-Tag, polyHistidine, Poly-histidine, polyhistidine tag | |
| This His Tag Antibody (RMH01) was developed against a mixture of a peptide with 6xHis-Tag at the N-terminus and another peptide with 6xHis Tag at the C-terminus. This antibody is a Rabbit-Human Chimeric antibody, made by fusion of the antigen binding region (variable domains of the heavy and light chains, VH and VL) of NBP2-61482, with the constant domain of Human IgG1. The antibody is recognized only by an anti-Human IgG secondary antibody. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot, Simple Western, Sandwich ELISA Capture, Sandwich ELISA Detection | |
| RMH01 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Tag | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction