missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody (H1alpha67) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-105JF585
This item is not returnable.
View return policy
Description
HIF-1 alpha Monoclonal antibody specifically detects HIF-1 alpha in Bovine,Canine,Feline,Ferret,Human,Monkey,Mouse,Porcine,Primate,Rabbit,Rat,Sheep,Xenopus samples. It is validated for ChIP assay,Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunoassay,Immunoblot,In vitro Assay,Gel Super Shift Assays,Proximity Ligation Assay,Tissue Culture Substratum,Ligand Activation,Immunohistochemistry Free-Floating,Simple Western,ChIP assay (ChIP),Gene Knock-Out,KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| HIF-1 alpha | |
| Monoclonal | |
| Janelia Fluor 585 | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
| This HIF-1 alpha Antibody (H1alpha67) was developed against a fusion protein containing amino acids 432 - 528 of human HIF-1 alpha [Uniprot# Q16665]. | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Cytokine Research, HIF Target Genes, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| 3091 | |
| Store at 4°C in the dark. | |
| IgG2b |
| ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Immunoblot, In vitro Assay, Gel Supershift, Proximity Ligation Assay, Tissue Culture Substratum, Ligand Activation, Immunohistochemistry (Free Floating), Simple Western, ChIP Assay, Gene Knock-Out, KnockDown, Immunocytochemistry/Immunofluorescence | |
| H1alpha67 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Feline, Ferret, Human, Monkey, Mouse, Pig, Primate, Rabbit, Rat, Sheep, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction