Learn More
Hexanoic anhydride, 97%, Thermo Scientific Chemicals
Catalog Number p-142561
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
2051-49-2
C12H22O3
214.3
MFCD00009509
PKHMTIRCAFTBDS-UHFFFAOYSA-N
hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
74918
hexanoyl hexanoate
CCCCCC(=O)OC(=O)CCCCC
2051-49-2
C12H22O3
214.3
MFCD00009509
PKHMTIRCAFTBDS-UHFFFAOYSA-N
hexanoic anhydride, caproic anhydride, hexanoic acid, anhydride, n-hexanoic anhydride, hexanoic acid, 1,1'-anhydride, unii-gg70y8ltby, n-caproic anhydride, caproic acid anhydride, capronic acid anhydride, n-hexanoic acid anhydride
74918
hexanoyl hexanoate
CCCCCC(=O)OC(=O)CCCCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -40°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 246°C to 248°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [CH3(CH2)4CO]2O
Refractive Index 1.4270 to 1.429
Quantity 50 g
Beilstein 02, 324
Specific Gravity 0.926
Solubility Information Solubility in water: 72g/L. Other solubilities: soluble in ethanol and ether
Formula Weight 214.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Hexanoic anhydride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Hexanoic anhydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.