missing translation for 'onlineSavingsMsg'
Learn More

Hexamethylacetone, 98%, Thermo Scientific Chemicals

Catalog Number p-7035737
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
815-24-7
C9H18O
142.242
MFCD00008845
UIQGEWJEWJMQSL-UHFFFAOYSA-N
13152
2,2,4,4-tetramethylpentan-3-one
CC(C)(C)C(=O)C(C)(C)C
This item is not returnable. View return policy
815-24-7
C9H18O
142.242
MFCD00008845
UIQGEWJEWJMQSL-UHFFFAOYSA-N
13152
2,2,4,4-tetramethylpentan-3-one
CC(C)(C)C(=O)C(C)(C)C

Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethylacetone is used to prepare di-tert.-butyladamantylcarbinol by reacting with 1-bromo-adamantane.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.824
Boiling Point 152°C to 153°C
Flash Point 34°C (93°F)
Refractive Index 1.419
Quantity 5 g
UN Number UN1224
Beilstein 1701122
Synonym hexamethylacetone, 2,2,4,4-tetramethyl-3-pentanone, di-tert-butyl ketone, 3-pentanone, 2,2,4,4-tetramethyl, 2,2,4,4-tetramethyl-pentan-3-one, 3-pentanone,2,2,4,4-tetramethyl, di-t-butyl ketone, acmc-20akrk, t-c4h9 2co, 2,4,4-tetramethyl-3-pentanone
Formula Weight 142.24
Percent Purity 98%
Chemical Name or Material Hexamethylacetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'