missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Hexafluoroglutaryl chloride, 97%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Specifications
Specifications
| Density | 1.64 |
| Boiling Point | 110°C to 112°C |
| Flash Point | None |
| Odor | Acrid |
| Refractive Index | 1.354 |
| Quantity | 1 g |
| UN Number | UN3265 |
| Beilstein | 1914561 |
| Sensitivity | Moisture sensitive |
| Formula Weight | 276.95 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction