missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexaammineruthenium(III) chloride, Ru 32.1% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
5 g
Description
Depending on the choice of suitable reductive or oxidative quencher this catalyst can be used to trigger photoreduction or photooxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Yellow |
Quantity | 0.25 g |
UN Number | UN3288 |
Solubility Information | Very soluble in water. |
IUPAC Name | ruthenium(3+) hexaamine trichloride |
Formula Weight | 309.61 |
Concentration or Composition (by Analyte or Components) | Ru 32.1% min. |
Physical Form | Powder |
Chemical Name or Material | Hexaammineruthenium(III) chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'