missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ HDPE Jerrican

Use these jerricans for liquid storage or transport. Buerkle™ HDPE Jerricans are UN transport approved and include a tamper-evident seal.

36.00 NOK - 266.00 NOK

Specifications

For Use With (Application) UN-Transport
Closure Type DIN45
Material HDPE
Certifications/Compliance UN approval 3H1/Y1.6/150
Shape Square
View More Specs

Products 5
Product Code Brand Dimensions (L x W x H) Capacity (Metric) Price Quantity & Availability  
Product Code Brand Dimensions (L x W x H) Capacity (Metric) Price Quantity & Availability  
10006791 View accessory products View Documents Buerkle™
1427-0010
230 x 190 x 310mm 10 L 69.45 NOK
Each View alternative products
Save Up to 
»
Estimated Shipment
09-01-2020
Add to basket
 
10770593 View accessory products View Documents Buerkle™
1427-0005
190 x 145 x 251mm 5 L 55.45 NOK
Each View alternative products
Save Up to 
»
Estimated Shipment
09-01-2020
Add to basket
 
10613693 View accessory products View Documents Buerkle™
1427-0020
290 x 246 x 385mm 20 L 185.00 NOK
Each View alternative products
Save Up to 
»
In Stock 1
Add to basket
 
11691819 View accessory products View Documents Buerkle™
1427-0030
320 x 284 x 435mm 30 L 266.00 NOK
Each View alternative products
Save Up to 
»
Estimated Shipment
15-01-2020
Add to basket
 
16339541 View Documents Buerkle™
1427-0002
153 x 115 x 208 mm 2.5 L 36.00 NOK
Each View alternative products
Save Up to 
»
Estimated Shipment
16-01-2020
Add to basket
 
Description

Description

  • HDPE
  • UN approval 3H1/X1.9/250
  • Screw lid with drain cock available as accessory
Specifications

Specifications

UN-Transport
HDPE
Square
DIN45
UN approval 3H1/Y1.6/150
Jerry Can