missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HDAC7 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | HDAC7 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884725
|
Proteintech
26207-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874725
|
Proteintech
26207-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Protein kinase D2 (PKD2) is a novel human serine threonine protein kinase gene. It exhibits the strongest homology to the serine threonine protein kinases PKD/PKCmu and PKCnu, particularly in the duplex zinc finger-like cysteine-rich motif, in the pleckstrin homology domain and in the protein kinase domain. In contrast, it shows only a low degree of sequence similarity to other members of the PKC family.Specifications
HDAC7 | |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q8WUI4 | |
51564 | |
HDAC7 Fusion Protein Ag24396 | |
Primary | |
-20°C | |
HDAC7 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
HD7, HD7A, HDAC7, HDAC7A, histone deacetylase 7, Histone deacetylase 7A | |
HDAC7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |