Learn More
Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
1130.00 NOK - 1735.00 NOK
Product Code | Brand | For Use With (Application) | No. of Tests | Range | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | No. of Tests | Range | Price | Quantity & Availability | |||
10060921
|
Hanna Instruments™
HI9373501 |
Total hardness medium range determination | 100 | 200 to 500mg/L as CaCO3 total hardness |
1130.00 NOK
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
11602690
|
Hanna Instruments™
HI9375601 |
Calcium Hardness determination | 100 | 0.00 to 2.70mg/L |
1735.00 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
10459202
|
Hanna Instruments™
HI93720-01 |
Calcium Hardness determination | 100 | 0.00 to 2.70mg/L calcium hardness |
1325.00 NOK
Each |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determination of various parameters of water hardness. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets are available for determination of:
- Calcium Hardness (using liquid or powdered reagents)
- Calcium and Magnesium Hardness (high range)
- Magnesium and Total Hardness
- Total Hardness (low, medium and/or high range)
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
1 Set | |
Water Testing Kit |