Learn More
Brand™ Haematocrit Sealing Compound
Quick-seals micro-haematocrit capillaries. BRAND™ Haematocrit Sealing Compoud is constructed of non-drying cement on a plastic plate. Up to 24 capillaries can be placed vertically in numbered positions along the sides of the tray.
Brand: Brand™ 749510
392.16 NOK valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Additional Details : Weight : 0.16560kg
Description
Non-drying cement on a plastic plate. For quick sealing of micro-haematocrit capillaries. Up to 24 capillaries can be placed vertically in numbered positions along the sides of the tray.Note: Hold the capillaries (∼ 2/3 full) near the empty end, and stick the empty end into the cement with a slight twist to seal it.
Specifications
Hematocrit Sealing Compound | |
Sealing of micro-haematocrit capillaries. | |
24 | |
10 Pk. |