Learn More
MedChemExpress γ-GT
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| CAS | 63699-78-5 |
| Solubility | H2O : ≥ 30 mg/mL (91.39 mM) |
| Content And Storage | 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Quantity | 100 mg |
| Molecular Weight (g/mol) | 328.28 g/mol |
| Product Type | Biochemical Assay Reagents |
| Purity or Quality Grade | 98.0% |
| Shipping Condition | Room temperature |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.