Learn More
Honeywell™ GRIP Latex Gloves
Click to view available options
Size:
9
10
Description
- Natural latex coated glove on cotton jersey base
- Crinkled finishing
- Knitted wrist
- The cotton absorbs perspiration thus provides greater comfort
- The ventilated back improves the glove's breathability
- The glove's crinkled latex coating provides resistance to abrasion and perforation as well as an excellent grip
- Natural latex provides excellent flexibility, synonymous with dexterity and comfort
- It also offers good abrasion and tear resistance
- Its crepe finish helps the user to grasp slippery parts
Specifications
Certifications/Compliance | EN 420-2003 Basic standard, EN 388-2016 Mechanical risks |
Coating Material | Latex |
Exterior Finish | Crinkle |
Material | Cotton |
Size | 10 |
Length (Metric) | 26 cm |