Learn More
Fisherbrand™ Gravity Convection Microbiological Incubator
Ideal incubator for basic incubation applications and heated storage
11261.33 NOK - 22016.53 NOK
Specifications
Convection Type Gravity
Display Digital
Material Stainless steel
No. of Shelves 2
Plug Type UK and EU
View More Specs
Products 3
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Exterior Price Quantity & Availability  
Product Code Brand Capacity (Metric) Dimensions (L x W x H) Exterior Price Quantity & Availability  
15895901 Fisherbrand™
75 L 11261.33 NOK
Each View alternative products
Save Up to 
»
Call for latest availability
+47 22 95 59 59
Add to basket
15835911 Fisherbrand™
194 L 22016.53 NOK
Each View alternative products
Save Up to 
»
In Stock 1
Add to basket
15815911 Fisherbrand™
117 L 15835.94 NOK
Each View alternative products
Save Up to 
»
Call for latest availability
+47 22 95 59 59
Add to basket
Description
Description
  • Ideal for microbiological incubation and heated storage
  • Maximum temperature 75°C
  • Single set point digital control
  • Inner glass door for minimal disturbance while viewing samples
  • Stackable, no tools or kit required
  • Access port standard
  • Stainless steel interior
  • Two year parts and labor warranty
  • CE marked
  • Two perforated stainless steel shelves included as standard
  • Extra shelves also available
Specifications
Specifications
Gravity
Stainless steel
UK and EU
Ambient +7° to 75°C
Digital
2
230 V, 50/60 Hz
2 Year, Parts and Labor