Learn More
Gramine, 99%, Thermo Scientific Chemicals
Catalog Number p-3758130
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
87-52-5
C11H14N2
174.25
MFCD00005629
OCDGBSUVYYVKQZ-UHFFFAOYSA-N
gramine, 3-dimethylaminomethyl indole, donaxine, donaxin, gramin, 1h-indole-3-methanamine, n,n-dimethyl, beta-dimethylaminomethylindole, 1-1h-indol-3-yl-n,n-dimethylmethanamine, indol-3-ylmethyldimethylamine, 1h-indol-3-ylmethyl dimethylamine
6890
CHEBI:28948
1-(1H-indol-3-yl)-N,N-dimethylmethanamine
CN(C)CC1=CNC2=CC=CC=C21
87-52-5
C11H14N2
174.25
MFCD00005629
OCDGBSUVYYVKQZ-UHFFFAOYSA-N
gramine, 3-dimethylaminomethyl indole, donaxine, donaxin, gramin, 1h-indole-3-methanamine, n,n-dimethyl, beta-dimethylaminomethylindole, 1-1h-indol-3-yl-n,n-dimethylmethanamine, indol-3-ylmethyldimethylamine, 1h-indol-3-ylmethyl dimethylamine
6890
CHEBI:28948
1-(1H-indol-3-yl)-N,N-dimethylmethanamine
CN(C)CC1=CNC2=CC=CC=C21
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 131.0°C to 139.0°C
Color Beige
Flash Point 167°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Merck Index 15, 4569
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether,toluene and chloroform, slightly soluble in cold acetone, practically insoluble in petr ether
Formula Weight 174.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Gramine
Show More Show Less
compliance-icons
Product Identifier
  • Gramine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.