Learn More
Fisherbrand™ Grade 144 Qualitative Phosphate-free Special Filter Paper
Fisherbrand™ Grade 144 Qualitative Phosphate-free Special Filter Paper are used for mineral analysis in soil extracts.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- Determination of K and P
Specifications
Folded Disc | |
3 to 5 μm | |
144 | |
0.16 mm | |
110 mm |
3 to 5 μm | |
Determination of K and P | |
100 Pack | |
110 mm |