missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 (Glycoprotein 2) / ZAP75 Mouse, Clone: GP2/1803, enQuireBio™
Mouse Monoclonal Antibody
2058.63 NOK - 5275.00 NOK
Specifications
Antigen | GP2 (Glycoprotein 2) / ZAP75 |
---|---|
Clone | GP2/1803 |
Concentration | 200μg/mL |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15940128
|
EnquireBio
2813-MSM3-P1 |
100 μg |
5275.00 NOK
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930128
|
EnquireBio
2813-MSM3-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
GP2 (Glycoprotein 2) / ZAP75 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human GP2 protein (apprx. aa 35-179) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
59kDa |
GP2/1803 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
RUO | |
2813 | |
IgG2a κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface and Secreted | |
PANC-1 Cells. Pancreas |