missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gossypol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Description
Gossypol acts as an inhibitor for several dehydrogenase enzymes. Gossypol has also long been known to possess antimalarial properties. Gossypol provided reliable contraception. gossypol provokes infertility and causes spermatogenesis. Gossypol is used as an active antifertility agent in a variety of male animals, including mice, rats, hamsters, monkeys, rabbits, and bulls.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Gossypol acts as an inhibitor for several dehydrogenase enzymes. Gossypol has also long been known to possess antimalarial properties. Gossypol provided reliable contraception. gossypol provokes infertility and causes spermatogenesis. Gossypol is used as an active antifertility agent in a variety of male animals, including mice, rats, hamsters, monkeys, rabbits, and bulls.
Solubility
Soluble in 100%ethanol (25 mg/ml), DMF (25 mg/ml), acetone, DMSO (25 mM), methanol (2 mg/ml), ether, chloroform, sodium carbonate, and dilute aqueous solutions of ammonia . Insoluble in water. Gossypol is a male antifertility agent with antispermatogenic activity and has been shown to contain antitumor, anitviral, and antioxidant properties.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Gossypol acts as an inhibitor for several dehydrogenase enzymes. Gossypol has also long been known to possess antimalarial properties. Gossypol provided reliable contraception. gossypol provokes infertility and causes spermatogenesis. Gossypol is used as an active antifertility agent in a variety of male animals, including mice, rats, hamsters, monkeys, rabbits, and bulls.
Solubility
Soluble in 100%ethanol (25 mg/ml), DMF (25 mg/ml), acetone, DMSO (25 mM), methanol (2 mg/ml), ether, chloroform, sodium carbonate, and dilute aqueous solutions of ammonia . Insoluble in water. Gossypol is a male antifertility agent with antispermatogenic activity and has been shown to contain antitumor, anitviral, and antioxidant properties.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Color | Yellow to Brown |
Quantity | 25 mg |
Merck Index | 14,4528 |
IUPAC Name | 7-(8-formyl-1,6,7-trihydroxy-3-methyl-5-propan-2-ylnaphthalen-2-yl)-2,3,8-trihydroxy-6-methyl-4-propan-2-ylnaphthalene-1-carbaldehyde |
Formula Weight | 518.56 |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Gossypol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'