Learn More
Corning™ Octagonal Polyethylene Terephtalate Bottles
Catalog Number 12557735
Click to view available options
Capacity:
16.9 oz., 500 mL
33.8 oz., 1000 mL
4.2 oz., 125 mL
8.45 oz., 250 mL
Closure Color:
Natural
Packaging:
Bulk
Tray Filmed Packed
Designed with a leak-resistant closure system
  • Break-resistant polyethylene terephthalate (PET) with transparency equivalent to glass
  • Molded graduations
  • Tamper-evident High-density Polyethylene (HDPE) cap
  • Film-packed with or without tray in folded outer bag
TRUSTED_SUSTAINABILITY
Capacity 16.9 oz., 500 mL
Closure Material HDPE
Closure Size 31 mm
Closure Type Screw Cap
Diameter (Metric) 76 mm
Height (English) 6.8 in.
Height (Metric) 173 mm
Capacity (Metric) 500 mL
Capacity (English) 16.9 oz.
Color Clear
Diameter (English) 2.9 in.
Diameter (English) Inner Neck 1.2 in.
Diameter (Metric) Inner Neck 31 mm
Material PET
Length (Metric) 74 mm
Packaging Tray Filmed Packed
Sterility Sterile
Width (Metric) 74 mm
Closure Color Natural
Graduations 50 to 500 mL
Certifications/Compliance ISO 9001:2008/ISO 11137 : STERILE SAL 10-3