missing translation for 'onlineSavingsMsg'
Learn More

Gomisin G, MedChemExpress

Product Code. p-200099542
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16489138

Brand: Medchem Express HYN0858

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gomisin G is an ethanolic extract of the stems of Kadsura interior; exhibits potent anti-HIV activity with EC50 and therapeutic index (TI) values of 0.006 microgram/mL and 300, respectively.

Gomisin G is an ethanolic extract of the stems of Kadsura interior

Specifications

Chemical Name or Material Gomisin G
CAS 62956-48-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H32O9
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (93.18 mM)
SMILES COC1=C(OCO2)C2=CC([C@H](OC(C3=CC=CC=C3)=O)[C@](O)(C)[C@@H](C)C4)=C1C(C4=CC(OC)=C5OC)=C5OC
Molecular Weight (g/mol) 536.57
Formula Weight 536.57
Percent Purity 99.9%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.