missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Antibody (gimsilumab) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28666JF635
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| GM-CSF | |
| Monoclonal | |
| Janelia Fluor 635 | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| CSF2/ GM-CSF | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 1437 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| gimsilumab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction