missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyphosate-13C2,15N, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Glyphosate 1,2-13C2 15 N |
| CAS | 1185107-63-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | 13C2 C H8 15 N O5 P |
| Quantity | 10 mg |
| Synonym | Glycine-1,2-13C2-15 N, N-(phosphonomethyl)- (ACI), N-(Phosphonomethyl)glycine-1,2-13C2-15 N (ACI), Glyphosate-13C2,15 N, Glyphosate 1,2-13C2 15 N, Glyphosate-13C2,15 N (glycine-13C2,15 N) |
| InChI Formula | InChI=1 S/C3H8NO5P/c5-3(6)1-4-2-10(7,8)9/h4H,1-2H2,(H,5,6)(H2,7,8,9)/i1+1,3+1,4+1 |
| SMILES | O[13 C](=O)[13 CH2][15 NH]CP(=O)(O)O |
| IUPAC Name | 2-(phosphonomethylamino)acetic acid |
| Molecular Weight (g/mol) | 172.05 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction