missing translation for 'onlineSavingsMsg'
Learn More

Glycylglycine, 0.2M buffer soln., pH 8.5, Thermo Scientific™

Catalog Number p-7041918
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
556-50-3
C4H8N2O3
132.119
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
11163
CHEBI:17201
2-[(2-aminoacetyl)amino]acetic acid
C(C(=O)NCC(=O)O)N
This item is not returnable. View return policy
556-50-3
C4H8N2O3
132.119
MFCD00008130
YMAWOPBAYDPSLA-UHFFFAOYSA-N
11163
CHEBI:17201
2-[(2-aminoacetyl)amino]acetic acid
C(C(=O)NCC(=O)O)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Concentration 0.2 M
pH 8.5
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Chemical Name or Material Glycylglycine buffer soln.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'