Learn More
Glycine benzyl ester hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Assay Percent Range | 97% |
Packaging | Glass Bottle |
Formula Weight | 201.65 |
Percent Purity | 97% |
Quantity | 1 g |
Chemical Name or Material | Glycine benzyl ester hydrochloride |