missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ultrapure, 99.5+%, Thermo Scientific Chemicals

Catalog Number p-7076625
Click to view available options
Quantity:
100 mL
250 mL
500 mL
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO
This item is not returnable. View return policy
56-81-5
C3H8O3
92.09
MFCD00004722
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
OCC(O)CO

Used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel’s sample well.

Glycerol, ultrapure is used in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel's sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component, glycerol is used as a solvent, plasticizer and humectant. Additional uses include as an aid in casting gradient gels, a protein stabilizer and storage buffer component.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 18°C
Density 1.26
Boiling Point 182°C (20 mmHg)
Flash Point 160°C (320°F)
Packaging Plastic bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.474
Quantity 100 mL
Beilstein 635685
Sensitivity Hygroscopic
Specific Gravity 1.26
Solubility Information Miscible with water
Formula Weight 92.09
Percent Purity ≥99.5%
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'