Learn More
Glufosinate-d3 Hydrochloride, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Glufosinate P-methyl-d3 hydrochloride |
| CAS | 1323254-05-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C5 2H3 H9 N O4 P . Cl H |
| Quantity | 10 mg |
| Synonym | 2-Amino-4-(hydroxy-(trideuteriomethyl)phosphoryl)butanoic acid hydrochloride, DL-Phosphinothricin-d3 hydrochloride, Glufosinate-d3 hydrochloride, Glufosinate-D3 (P-methyl-D3) hydrochloride, (2 RS)-2-Amino-4-(hydroxy(methyl-d3)phosphinyl)butanoic acid hydrochloride, 2-Amino-4-[hydroxy(methyl-d3)phosphoryl]butanoic acid hydrochloride |
| InChI Formula | InChI=1 S/C5H12NO4P.ClH/c1-11(9,10)3-2-4(6)5(7)8;/h4H,2-3,6H2,1H3,(H,7,8)(H,9,10);1 H/i1D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])P(=O)(O)CCC(N)C(=O)O |
| IUPAC Name | 2-amino-4-[hydroxy(trideuteriomethyl)phosphoryl]butanoic acid;hydrochloride |
| Molecular Weight (g/mol) | 220.61 |
| Show More |