Learn More
GLIPR1L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20828-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850415
  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
Description
The function of this protein remains unknown.
Specifications
Specifications
GLIPR1L1
Polyclonal
Unconjugated
GLIPR1L1
ALKN2972, GLIPR1 like protein 1, GLIPR1L1, PRO7434
Rabbit
Antigen Affinity Chromatography
RUO
256710
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6UWM5
GLIPR1L1
GLIPR1L1 Fusion Protein Ag14852