Learn More
Glassy carbon splinter powder, 20 to 50μm, type 1, Alfa Aesar™
Used as such in purifying metals, in electrodes, electrical devices & steel.
Brand: Alfa Aesar 041258.30
Additional Details : CAS Number : 7440-44-0 Weight : 0.25000kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III
Quantity | 250g |
---|
Chemical Identifiers
MFCD00133992 |
Specifications
Glassy carbon splinter powder | |
250g | |
UN1325 |
Glass bottle | |
MFCD00133992 | |
Insoluble in water. |