Learn More
Gibco™ GM-CSF Recombinant Rat Protein
Recombinant GM-CSF is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method Exchange Chromatography
Endotoxin Concentration <0.1ng/μg
Gene Alias GM-CSF
Gene ID (Entrez) 116630
View More Specs
Description
Description
Recombinant rat GM-CSF is intended for use in cell culture applications. GM-CSF is involved in many biological responses including the growth and development of granulocyte and macrophage progenitor cells, stimulation and the initiation of differentiation of myeloblasts and monoblasts, and chemotaxis of eosinophils.
Shipping Condition: Wet Ice.
Specifications
Specifications
Exchange Chromatography
<0.1ng/μg
GM-CSF
116630