missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems GFP Fusion Human Mesothelin / MSLN (296-580) Protein, His Tag
5297.56 NOK - 21038.87 NOK
Specifications
Accession Number | AAH09272.1 |
---|---|
Molecular Weight (g/mol) | 60.4 kDa |
Source | GFP Fusion Human Mesothelin (296-580), His Tag (MSN-HG2P4) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1). |
Immunogen | Glu 296 - Gly 580 |
Storage Requirements | -20°C, avoid light |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16437889
|
ACROBiosystems
MSN-HG2P4-200UG |
200 μg |
21038.87 NOK
200µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16447889
|
ACROBiosystems
MSN-HG2P4-25UG |
25 μg |
5297.56 NOK
25µg |
Estimated Shipment: 10-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mesothelin (MSLN) is also known as CAK1 antigen, Pre-pro-megakaryocyte-potentiating factor, which belongs to the mesothelin family. Mesothelin / MSLN can be proteolytically cleaved into the following two chains by a furin-like convertase: Megakaryocyte-potentiating factor (MPF) and the cleaved form of mesothelin. Both MPF and the cleaved form of mesothelin are N-glycosylated. Mesothelin / MSLN can interacts with MUC16. The membrane-anchored forms of MSLN may play a role in cellular adhesion. MPF potentiates megakaryocyte colony formation in vitro.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
AAH09272.1 | |
GFP Fusion Human Mesothelin (296-580), His Tag (MSN-HG2P4) is expressed from human 293 cells (HEK293). It contains AA Glu 296 - Gly 580 (Accession # AAH09272.1). | |
-20°C, avoid light | |
Mesothelin | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Fluorescent labeled protein |
60.4 kDa | |
Glu 296 - Gly 580 | |
MSLN,Mesothelin,MPF | |
GFP Fusion Protein | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |