Learn More
Geranyl bromide, 96%
Product Code. 11488036
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11488036
Brand: Thermo Scientific Alfa Aesar A14093.14
Request Bulk or Custom Format
CAS: 6138-90-5 | C10H17Br | 217.15 g/mol
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.
Solubility
Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6138-90-5
Molecular Formula C10H17Br
Molecular Weight (g/mol) 217.15
MDL Number MFCD00000243
InChI Key JSCUZAYKVZXKQE-JXMROGBWSA-N
Synonym geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
PubChem CID 5365867
IUPAC Name (2E)-1-bromo-3,7-dimethylocta-2,6-diene
SMILES CC(=CCCC(=CCBr)C)C
Specifications
Density 1.094
Boiling Point 99°C to 101°C (10 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.504
Quantity 25 g
Beilstein 1422412
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 217.16
Percent Purity 96%
Chemical Name or Material Geranyl bromide