missing translation for 'onlineSavingsMsg'
Learn More

S Murray™ General-Purpose Narrow-Neck Bottles

Catalog Number p-8000417
Click to view available options
Capacity (Metric):
10,000 mL
5000 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Use these bottles for general storage. S Murray™ General-Purpose Narrow-Neck Bottles feature molded graduations and s covered swivel handle.

  • High density polyethylene, octagonal bottles
  • With screw cap
Closure Type Screw Cap
Capacity (Metric) 5000 mL
Color Natural
For Use With (Application) For storage and transportation of liquids
Material MDPE
Closure Included Yes
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'