Learn More
GALNT5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552635
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AERDLNVTIS LSTDRPKQRS QAVANERAHP ASTAVPKSGE AMALNKTKTQ SKEVNANKHK ANTSLPFPKF TVNSNRLRKQ SINETPLGSL SKDDGARGAH GKKLNFSESH LVIITKEEEQ KADPKEVSNS KTKTIFPKVL GKS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 53%.
GALNT5 Polyclonal antibody specifically detects GALNT5 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
GALNT5 | |
Polyclonal | |
Unconjugated | |
GALNT5 | |
GALNACT5; GalNAc-T5; GALNT5; Polypeptide GalNAc transferase 5; polypeptide N-acetylgalactosaminyltransferase 5; pp-GaNTase 5; protein-UDP acetylgalactosaminyltransferase 5; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 5; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5 (GalNAc-T5) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11227 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z7M9 | |
GALNT5 | |
Recombinant protein corresponding to Human GALNT5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only