missing translation for 'onlineSavingsMsg'
Learn More

Galectin-3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen Galectin-3
Concentration 0.27 mg/mL
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16803934
View Documents Promotion Details
Proteintech
14979-1-AP-20UL
20 μL
1611.84 NOK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16893924
View Documents Promotion Details
Proteintech
14979-1-AP-150UL
150 μL
4593.76 NOK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The monoclonal antibody B2C10 reacts with galectin-3, a 30 kDa protein. Galectin-3 is a member of the galectin family. The protein is compose of three domains: a small amino-terminal domain, a carboxyl-terminal carbohydrate recognition domain (CRD) and amino-terminal domain containing repeating elements. Galectin-3 is normally distributed in epithelia of many organs and various inflammatory cells, including macrophages, as well as dendritic cells and Kupffer cells. The expression of this lectin is up-regulated during inflammation, cell proliferation, cell differentiation and through trans-activation by viral proteins. The expression is also affected by neoplastic transformation: up-regulated in certain types of lymphomas and thyroid carcinoma, while down-regulated in other types of maligcies, such as colon, breast, ovarian and uterine carcinomas.
Specifications

Specifications

Galectin-3
Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Human, Rat
P08699, P16110, P17931
16854, 3958, 83781
Galectin-3 Fusion Protein Ag6891
Primary
-20°C
LGALS3
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
35 kDa lectin, CBP 35, CBP35, Gal 3, GAL3, Galactose specific lectin 3, Galactoside binding protein, GALBP, Galectin 3, Galectin-3, GALIG, IgE binding protein, L 31, Laminin binding protein, Lectin L 29, LGALS2, LGALS3, Mac 2 antigen, MAC2
Lgals3
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications