Learn More
Gabapentin Methyl Ester Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Gabapentin Methyl Ester Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Gabapentin Methyl Ester Hydrochloride |
| CAS | 63562-01-6 |
| Analyte or Component Names | Gabapentin Methyl Ester Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H19 N O2 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-(Aminomethyl)-cyclohexaneacetic acid methyl ester hydrochloride (1:1), Cyclohexaneacetic acid, 1-(aminomethyl)-, methyl ester, hydrochloride (9CI), (1-Aminomethylcyclohexyl)acetic acid methyl ester hydrochloride |
| InChI Formula | InChI=1S/C10H19NO2.ClH/c1-13-9(12)7-10(8-11)5-3-2-4-6-10;/h2-8,11H2,1H3;1H |
| SMILES | Cl.COC(=O)CC1(CN)CCCCC1 |
| IUPAC Name | methyl 2-[1-(aminomethyl)cyclohexyl]acetate;hydrochloride |
| Show More |