Learn More
Furan-d4, CDN
Product Code. 30321007
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321007
Brand: CDN CDND01845G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Furan D4
CAS 6142-90-1
Molecular Formula C4 2H4 O
Quantity 5 g
Synonym Furan-2,3,4,5-d4, Furan-d4 (7 CI,8 CI,9 CI), Perdeuteriofuran, Tetradeuteriofuran
InChI Formula InChI=1 S/C4H4O/c1-2-4-5-3-1/h1-4 H/i1D,2 D,3 D,4 D
SMILES [2 H]c1oc([2 H])c([2 H])c1[2 H]
IUPAC Name 2,3,4,5-tetradeuteriofuran
Molecular Weight (g/mol) 72.1
Formula Weight 72.0513 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity