Learn More
DWK Life Sciences DURAN™ Blank for separating and dropping funnel, cylindrical with stopper bed for ISO 4800
Raw body for further processing as dropping funnel.
Brand:  DWK Life Sciences 210143606
2180.25 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 1.55000kg
 View more versions of this product
Product Code. 15241629
  • 2295.00 NOK / Pack of 10
Estimated Shipment: 14-05-2024
Description
DURAN™ dropping funnels are used, for example, for the metered supply of liquid to a reaction mixture.
  • Cylindrical with stopper bed for ISO 4800
  • Glass articles for further processing
Specifications
Specifications
52 mm
Boro 3.3
Blank for further processing as dropping funnel; Phase separation
250 mL
Yes
10/Pk.