Learn More
Thermo Scientific™ Frosted Microscope Slides, Cut
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Brand:  Thermo Scientific™ AAAC0600075052EB0MNZ#0#G#
Additional Details : Weight : 0.51000kg
 View more versions of this product
Product Code. 15241895
  • 5290.00 NOK / Pack of 100
Call for latest availability
+47 22 95 59 59
Add to basket
Description
Description
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specifications
Specifications
75mm
1mm
Uncoated
Soda-lime glass
Yes
60mm
N/A
5mm
100 pieces in paper
75 x 60mm