Learn More
VACUUBRAND™ FPM O-rings
30.65 NOK - 43.95 NOK
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
10000512
|
BrandTech™ VACUUBRAND™
23120553 |
28 x 2.5mm |
37.20 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
10010522
|
VACUUBRAND™
23120790 |
13 x 2mm |
31.90 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11919781
|
VACUUBRAND™
20637090 |
28 x 1.5mm |
30.65 NOK
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
11900692
|
VACUUBRAND™
20638138 |
37 x 1.5mm |
43.95 NOK
Each |
Estimated Shipment: 24-05-2024
Log in to see stock. |
|||||
11939731
|
VACUUBRAND™
20637978 |
5 x 3mm |
43.95 NOK
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
11979731
|
VACUUBRAND™
20638511 |
16 x 2mm |
30.65 NOK
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Specifications
O-Ring | |
FPM | |
1Pack |