missing translation for 'onlineSavingsMsg'
Learn More

Formamide, 99%, Thermo Scientific Chemicals

Catalog Number p-7081205
Click to view available options
Quantity:
500 g
2500 g
10 kg
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O
This item is not returnable. View return policy
75-12-7
CH3NO
45.04
MFCD00007941
ZHNUHDYFZUAESO-UHFFFAOYSA-N
methanamide, carbamaldehyde, formimidic acid, formic acid, amide, formamid, methanoic acid, amide, amid kyseliny mravenci, amid kyseliny mravenci czech, formic amide, hsdb 88
713
CHEBI:48431
formamide
NC=O

Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assembly of polymer nanofilms. It is used in the preparation of brain sections of mice for BrdU immunohistochemistry and fish analysis of HeLa cells. It essentially plays a vital role in crystallization of pharmaceuticals, in soil stabilization, as a solvent in inks, as a component of liquid fertilizers, as a heat-resistant coatings and in some personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Formamide is used for cryopreservation of tissues and organs and in hybridization buffers. It acts as an RNA stabiliser in gel electrophoresis by deionizing RNA. It is used as an alternative solvent for the electrostatic self-assemb
TRUSTED_SUSTAINABILITY
Melting Point 2°C
Density 1.134
Boiling Point 210°C
Quantity 500 g
Flash Point 150°C (302°F)
Odor Ammonia-like
Refractive Index 1.4475
Linear Formula HCONH2
Beilstein 505995
Merck Index 14,4237
Solubility Information It is miscible with water.
Formula Weight 45.04
Percent Purity 99%
Chemical Name or Material Formamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'