missing translation for 'onlineSavingsMsg'
Learn More

FNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 19847-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889324

  • 3468.15 NOK / 150µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FNIP1 encodes a protein that binds to the tumor suppressor protein folliculin and to AMP-activated protein kinase (AMPK). The encoded protein participates in the regulation of cellular metabolism and nutrient sensing by modulating the AMPK and target of rapamycin signaling pathways. This gene has a closely related paralog that encodes a protein with similar binding activities. Both related proteins also associate with the molecular chaperone heat shock protein-90 (Hsp90) and negatively regulate its ATPase activity and facilitate its association with folliculin. Diseases associated with FNIP1 include Birt-Hogg-Dube Syndrome and Pleural Disease.
Specifications

Specifications

FNIP1
Polyclonal
Unconjugated
FNIP1
FNIP1, KIAA1961
Rabbit
Antigen Affinity Chromatography
RUO
100362347, 216742, 96459
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FD7, Q8TF40
FNIP1
FNIP1 Fusion Protein Ag13899
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers