missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluoride Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, ready to use, Fisher Chemical™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
500 mL
Description
Fluoride Standard Solution 1000ppm (1mg/mL), for Ion Pair Chromatography, ready to use
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Color | Colorless |
Vapor Pressure | 14mm Hg at 20°C deg C |
Boiling Point | 100°C |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'