Learn More
BioTek Instruments Fluorescence Filter
Fluorescence Filter for use with BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Brand:  BioTek Instruments 7082221
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12505506
  • 4475.00 NOK / Each
Estimated Shipment: 10-12-2021
Description
Description
Fluorescence Filter for use with BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Specifications
Specifications
485/20 nm
Cytation 3, Cytation 5, Cytation Multi-Mode, FLx800, Synergy 2, Synergy H1, Synergy HTX Multi-Mode Readers
Fluorescence Filter