Learn More
BioTek Instruments Fluorescence Filter
Fluorescence Filter for use with BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Brand:  BioTek Instruments 7082251
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12552787
  • 5721.00 NOK / Each
Estimated Shipment: 15-12-2021
Description
Description
Fluorescence Filter for use with BioTek Microplate readers and imagers: Cytation 5, Synergy H1, Synergy HTX and Synergy LX
Specifications
Specifications
620/15 nm
Cytation 3, Cytation 5, FLx800, Synergy 2, Synergy H1, Synergy HTX
Fluorescence Filter