missing translation for 'onlineSavingsMsg'
Learn More

Fluorene-1-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-7027944
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6276-03-5
C14H10O2
210.23
MFCD00001130
HTPXFGUCAUTOEL-UHFFFAOYSA-N
1-fluorenecarboxylic acid, fluorene-1-carboxylic acid, fluorenecarboxylic acid, unii-ao792vd660, 1-fluorene carboxylic acid, 9h-fluorene-1-carboxylicacid, htpxfgucautoel-uhfffaoysa
80468
9H-fluorene-1-carboxylic acid
C1C2=CC=CC=C2C3=C1C(=CC=C3)C(=O)O
This item is not returnable. View return policy
6276-03-5
C14H10O2
210.23
MFCD00001130
HTPXFGUCAUTOEL-UHFFFAOYSA-N
1-fluorenecarboxylic acid, fluorene-1-carboxylic acid, fluorenecarboxylic acid, unii-ao792vd660, 1-fluorene carboxylic acid, 9h-fluorene-1-carboxylicacid, htpxfgucautoel-uhfffaoysa
80468
9H-fluorene-1-carboxylic acid
C1C2=CC=CC=C2C3=C1C(=CC=C3)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 246°C to 248°C
Odor Odorless
Quantity 1 g
Formula Weight 210.23
Percent Purity 97%
Chemical Name or Material Fluorene-1-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'