Learn More
Cole-Parmer™ Stuart™ Florentine Flask For Stuart™ Rotary Evaporators
Evaporating flask used with Stuart™ Rotary Evaporators.
Brand:  Cole-Parmer™ FD250/4RE/1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12497732
  • 1277.86 NOK / Each
Estimated Shipment: 29-10-2021
Add to basket
Description
Description
  • For use with Stuart™ rotary evaporators
  • Flat bottom
  • 29/32 socket
Specifications
Specifications
Evaporating flask
Glass
Flat
250 mL
29/32