missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLJ14213 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | FLJ14213 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884845
|
Proteintech
26355-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16874845
|
Proteintech
26355-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Proline-rich protein 5-like (PRR5L), also known as Protor-2, is structurally similar to PRR5 and also binds to the mTOR complex 2 (mTORC2). PRR5L is not required for mTORC2 integrity or kinase activity, but dissociates from mTORC2 upon knockdown of tuberous sclerosis complex 1 (TSC1) and TSC2. Knockdown of PRR5L reduced apoptosis in mTORC2 deficient cells as well as cells where mTOR was active, suggesting that mTORC2-dissociated PRR5L may promote apoptosis, and thus play a role in the balance between cell growth and cell death.Specifications
FLJ14213 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q6MZQ0 | |
79899 | |
FLJ14213 Fusion Protein Ag23837 | |
Primary | |
-20°C | |
PRR5L |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FLJ14213, Proline rich protein 5 like, Protein observed with Rictor 2, protor 2, PROTOR2, PRR5L | |
PRR5L | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |