Learn More
Memmert™ Flexible Pt100 For Chamber Or Load, Display And Atmocontrol Compatible, For U30
Flexible temperature sensor for positioning in chamber or in load. Memmert™ PT100 Temperature Sensor is designed with a 4-pin connector for external temperature recording.
Brand: Memmert™ U30H8
Additional Details : Weight : 0.01100kg
Includes: 4-Pin Connector for Temperature Recording
Description
- Can be positioned in chamber or in load
- Includes 4-pin connector, according to NAMUR NE 28, for external temperature recording (load temperature) max. 3 sensors
- For use in Memmert™ universal ovens
Specifications
Secondary/Additional PT 100 Sensor (up to 3 can be attached in total) | |
Models VO, VOcool, HCP, TTC, CTC, U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH, 108 | |
Temperature Sensor | |
Load/Chamber Temperature Measurement |